Gene Transcripts

Gene Symbol
Ltv1
Entrez Gene ID
361452
Gene Name
LTV1 ribosome biogenesis factor
Gene Aliases
RGD1311264
Chromosome Location
Chr. 1: 7323043 - 7335766 on Build Rnor_6.0
UniGene ID
Rn.7361
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014157.1 NP_001014179.1 10, 11 1329
GenBank mRNA BC079397.1 - 10, 11 1329
FQ213117.1 - 5, 6 765

View Full Product Details