Gene Transcripts

Gene Symbol
Ppme1
Entrez Gene ID
361613
Gene Name
protein phosphatase methylesterase 1
Gene Aliases
PME-1, RGD1309683
Chromosome Location
Chr. 1: 165335242 - 165382216 on Build Rnor_6.0
UniGene ID
Rn.137659
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191838.1 NP_001178767.1 2, 3 342

View Full Product Details