Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006233886.2 | XP_006233948.1 | 11 | 5607 | |
XM_006233887.2 | XP_006233949.1 | 12 | 5610 | ||
XM_006233888.2 | XP_006233950.1 | 11 | 5606 | ||
XM_008761681.1 | XP_008759903.1 | 11 | 5721 | ||
XM_008761682.1 | XP_008759904.1 | 8 | 4975 | ||
GenBank mRNA | FQ222225.1 | - | 2 | 425 |