Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191857.1 | NP_001178786.1 | 21 | 2018 | |
XM_006235772.2 | XP_006235834.1 | 21 | 2074 | ||
XM_006235773.2 | XP_006235835.1 | 21 | 2483 | ||
XM_006235774.2 | XP_006235836.1 | 21 | 2074 | ||
XM_006235775.2 | XP_006235837.1 | 21 | 2074 | ||
XM_006235776.2 | XP_006235838.1 | 21 | 2074 | ||
XM_006235778.2 | XP_006235840.1 | 14 | 1613 |