Gene Transcripts

Gene Symbol
KSR1
Entrez Gene ID
8844
Gene Name
kinase suppressor of ras 1
Gene Aliases
KSR, RSU2
Chromosome Location
Chr. 17: 27456441 - 27626438 on Build GRCh38
UniGene ID
Hs.133534
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014238.1 NP_055053.1 8 1150
XM_006722154.3 XP_006722217.1 7 1302
XM_011525429.2 XP_011523731.1 7 1302
XM_011525430.2 XP_011523732.1 7 1304
XM_011525431.2 XP_011523733.1 7 1304
XM_017025267.1 XP_016880756.1 7 1302
XM_017025268.1 XP_016880757.1 7 1302
XM_017025269.1 XP_016880758.1 7 1302
XM_017025270.1 XP_016880759.1 6 1037
XM_017025271.1 XP_016880760.1 8 2009
XM_017025272.1 XP_016880761.1 8 1992
XM_017025273.1 XP_016880762.1 7 1815
XM_017025274.1 XP_016880763.1 7 1018
XM_017025275.1 XP_016880764.1 7 1005
XM_017025276.1 XP_016880765.1 8 1957
XM_017025277.1 XP_016880766.1 3 700
XM_017025278.1 XP_016880767.1 3 710
XM_017025279.1 XP_016880768.1 7 1319
XM_017025280.1 XP_016880769.1 7 1319
GenBank mRNA BC035488.1 - 8 2128
BC042106.1 - 8 1149
U43586.1 - 4 390

View Full Product Details