Gene Transcripts

Gene Symbol
Mios
Entrez Gene ID
362324
Gene Name
missing oocyte, meiosis regulator, homolog (Drosophila)
Gene Aliases
RGD1308432
Chromosome Location
Chr. 4: 34238381 - 34264015 on Build Rnor_6.0
UniGene ID
Rn.103328
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173554.1 NP_001167025.1 3 1312
XM_006236090.2 XP_006236152.1 2 1312

View Full Product Details