Gene Transcripts

Gene Symbol
Tctex1d1
Entrez Gene ID
362553
Gene Name
Tctex1 domain containing 1
Gene Aliases
RGD1311932
Chromosome Location
Chr. 5: 122498884 - 122522110 on Build Rnor_6.0
UniGene ID
Rn.206137
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135030.1 NP_001128502.1 5 688
XM_006238459.2 XP_006238521.1 5 1444
XM_008763888.1 XP_008762110.1 4 515
XM_008763889.1 XP_008762111.1 5 3361
XM_008763890.1 XP_008762112.1 4 1222
XM_008763891.1 XP_008762113.1 4 665

View Full Product Details