Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004665.4 | NP_004656.2 | 4 | 792 | |
NM_078488.2 | NP_511043.1 | 5 | 620 | ||
NR_110143.1 | 4 | 792 | |||
XM_006715593.3 | XP_006715656.1 | 6 | 767 | ||
XM_011536231.1 | XP_011534533.1 | 4 | 688 | ||
XM_017011407.1 | XP_016866896.1 | 5 | 1403 | ||
XM_017011408.1 | XP_016866897.1 | 6 | 772 | ||
XM_017011409.1 | XP_016866898.1 | 6 | 665 | ||
GenBank mRNA | AB026705.1 | - | 4 | 674 | |
AJ132100.1 | - | 4 | 678 | ||
AK290908.1 | - | 4 | 688 | ||
AK292125.1 | - | 5 | 608 | ||
BC126145.1 | - | 4 | 754 | 8991861 | |
BC126147.1 | - | 4 | 754 | 8991863 | |
D89974.1 | - | 4 | 678 |