Gene Transcripts

Gene Symbol
Ctrc
Entrez Gene ID
362653
Gene Name
chymotrypsin C (caldecrin)
Gene Aliases
Chromosome Location
Chr. 5: 160394677 - 160403373 on Build Rnor_6.0
UniGene ID
Rn.19214
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077649.2 NP_001071117.1 4 337
GenBank mRNA S80379.1 - 4 338

View Full Product Details