Gene Transcripts
- Gene Symbol
- Ccdc88c
- Entrez Gene ID
- 362770
- Gene Name
- coiled-coil domain containing 88C
- Gene Aliases
- RGD1307429
- Chromosome Location
- Chr. 6: 124905811 - 125028013 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_343096.6 |
XP_343097.4
|
30 |
6068 |
|
View Full Product Details