Gene Transcripts

Gene Symbol
R3hdm2
Entrez Gene ID
362894
Gene Name
R3H domain containing 2
Gene Aliases
RGD1310066
Chromosome Location
Chr. 7: 70676795 - 70805255 on Build Rnor_6.0
UniGene ID
Rn.203556
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130557.1 NP_001124029.1 2 218
XM_006241469.2 XP_006241531.1 4 1303
XM_006241470.2 XP_006241532.1 3 1159
XM_006241471.2 XP_006241533.1 4 1269
XM_006241472.2 XP_006241534.1 2 1067
XM_006241473.2 XP_006241535.1 4 1208
XM_006241475.2 XP_006241537.1 4 1303
XM_006241476.2 XP_006241538.1 4 1303
XM_006241477.2 XP_006241539.1 4 1303
XM_006241478.2 XP_006241540.1 4 1303
XM_006241479.2 XP_006241541.1 4 1303
XM_006241480.2 XP_006241542.1 4 1303
XM_006241481.2 XP_006241543.1 4 1302
XM_006241482.2 XP_006241544.1 4 1302
XM_006241483.2 XP_006241545.1 3 1158
XM_006241484.2 XP_006241546.1 4 1302
XM_008765408.1 XP_008763630.1 5 614
XM_008765409.1 XP_008763631.1 5 715
XM_008765410.1 XP_008763632.1 5 482
XM_008765411.1 XP_008763633.1 4 401
XM_008765412.1 XP_008763634.1 4 511
XM_008765413.1 XP_008763635.1 3 415

View Full Product Details