Gene Transcripts

Gene Symbol
Dalrd3
Entrez Gene ID
363146
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
Chromosome Location
Chr. 8: 117355943 - 117358819 on Build Rnor_6.0
UniGene ID
Rn.120770
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014214.1 NP_001014236.1 9 1244
GenBank mRNA BC082046.1 - 9 1244

View Full Product Details