Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005407.1 | NP_001005407.1 | 26 | 5143 | |
NM_021098.2 | NP_066921.2 | 27 | 5161 | ||
XM_005255652.4 | XP_005255709.1 | 26 | 5022 | ||
XM_006720963.3 | XP_006721026.1 | 26 | 5022 | ||
XM_006720964.3 | XP_006721027.1 | 27 | 5007 | ||
XM_006720965.3 | XP_006721028.1 | 26 | 4989 | ||
XM_006720967.3 | XP_006721030.1 | 27 | 4946 | ||
XM_006720968.3 | XP_006721031.1 | 26 | 4928 | ||
XM_011522724.2 | XP_011521026.1 | 23 | 4471 | ||
XM_017023819.1 | XP_016879308.1 | 26 | 4983 | ||
XM_017023820.1 | XP_016879309.1 | 27 | 5084 | ||
XM_017023821.1 | XP_016879310.1 | 27 | 5084 | ||
GenBank mRNA | AF051946.3 | - | 27 | 4992 | |
AF073931.1 | - | 27 | 5161 | ||
AJ420779.1 | - | 25 | 4895 | ||
AK074965.1 | - | 4 | 544 | ||
DQ363526.1 | - | 9 | 1119 | ||
DQ363527.1 | - | 9 | 1119 | ||
DQ363528.1 | - | 10 | 1137 | ||
DQ363529.1 | - | 9 | 1152 |