Gene Transcripts

Gene Symbol
Cnnm4
Entrez Gene ID
363216
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
Chromosome Location
Chr. 9: 43049587 - 43088690 on Build Rnor_6.0
UniGene ID
Rn.857
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271050.1 NP_001257979.1 1 995
XM_006244827.2 XP_006244889.1 1 941

View Full Product Details