Gene Transcripts

Gene Symbol
Ms4a15
Entrez Gene ID
365408
Gene Name
membrane-spanning 4-domains, subfamily A, member 15
Gene Aliases
RGD1561158
Chromosome Location
Chr. 1: 227033796 - 227048718 on Build Rnor_6.0
UniGene ID
Rn.231430
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191897.1 NP_001178826.1 2 221
XM_006231063.2 XP_006231125.1 2 1713
XM_008760241.1 XP_008758463.1 2 1727

View Full Product Details