Gene Transcripts

Gene Symbol
CLDN8
Entrez Gene ID
9073
Gene Name
claudin 8
Gene Aliases
HEL-S-79
Chromosome Location
Chr. 21: 30214006 - 30216151 on Build GRCh38
UniGene ID
Hs.162209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199328.2 NP_955360.1 1 837
GenBank mRNA AK022269.1 - 1 760
AK223512.1 - 1 731
AY358707.1 - 1 707
BC020866.1 - 1 720 4594155
BC058004.1 - 1 686 4606880
GQ472207.1 - 1 720

View Full Product Details