Gene Transcripts

Gene Symbol
Spink14
Entrez Gene ID
408231
Gene Name
serine peptidase inhibitor, Kazal type 14
Gene Aliases
Spink5l2
Chromosome Location
Chr. 1: 229349133 - 229351168 on Build Rnor_6.0
UniGene ID
Rn.142547
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008875.1 NP_001008875.1 3 211

View Full Product Details