Gene Transcripts

Gene Symbol
Ly49si1
Entrez Gene ID
494206
Gene Name
immunoreceptor Ly49si1
Gene Aliases
Chromosome Location
Chr. 4: 163736176 - 163851595 on Build Rnor_6.0
UniGene ID
Rn.129033
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009497.1 NP_001009497.1 6 730
GenBank mRNA AY659935.1 - 6 730

View Full Product Details