Gene Transcripts

Gene Symbol
Vom1r37
Entrez Gene ID
494272
Gene Name
vomeronasal 1 receptor 37
Gene Aliases
V1rd6
Chromosome Location
Chr. 1: 72210037 - 72210999 on Build Rnor_6.0
UniGene ID
Rn.129428
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008923.1 NP_001008923.1 1 913
GenBank mRNA AY510333.1 - 1 913

View Full Product Details