Gene Transcripts

Gene Symbol
Coq2
Entrez Gene ID
498332
Gene Name
coenzyme Q2 4-hydroxybenzoate polyprenyltransferase
Gene Aliases
RGD1306722
Chromosome Location
Chr. 14: 10581104 - 10601093 on Build Rnor_6.0
UniGene ID
Rn.7463
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044255.1 NP_001037720.1 2 388
XM_006250675.2 XP_006250737.1 2 420
GenBank mRNA BC099827.1 - 2 388

View Full Product Details