Gene Transcripts

Gene Symbol
Nrg3
Entrez Gene ID
498596
Gene Name
neuregulin 3
Gene Aliases
RGD1559678
Chromosome Location
Chr. 16: 15763876 - 15818330 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771178.1 XP_008769400.1 5 1023

View Full Product Details