Gene Transcripts

Gene Symbol
Idnk
Entrez Gene ID
498695
Gene Name
idnK, gluconokinase homolog (E. coli)
Gene Aliases
Chromosome Location
Chr. 17: 6885323 - 6892620 on Build Rnor_6.0
UniGene ID
Rn.207173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037362.1 NP_001032439.1 4 207
XM_006253563.2 XP_006253625.1 3 338
GenBank mRNA BC107923.1 - 4 207

View Full Product Details