Gene Transcripts

Gene Symbol
Zfp61
Entrez Gene ID
499094
Gene Name
zinc finger protein 61
Gene Aliases
Chromosome Location
Chr. 1: 81122789 - 81130820 on Build Rnor_6.0
UniGene ID
Rn.122263
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017512.2 NP_001017512.2 6 931
XM_008758962.1 XP_008757184.1 7 1313
GenBank mRNA BC079015.1 - 6 878
CV078078.1 - 1 204

View Full Product Details