Gene Transcripts

Gene Symbol
Calhm3
Entrez Gene ID
499368
Gene Name
calcium homeostasis modulator 3
Gene Aliases
Fam26a, RGD1560215
Chromosome Location
Chr. 1: 266929299 - 266934303 on Build Rnor_6.0
UniGene ID
Rn.228197
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191956.1 NP_001178885.1 3 658

View Full Product Details