Gene Transcripts

Gene Symbol
Gstm6
Entrez Gene ID
499688
Gene Name
glutathione S-transferase, mu 6
Gene Aliases
RGD1563391
Chromosome Location
Chr. 2: 210733594 - 210738368 on Build Rnor_6.0
UniGene ID
Rn.133165
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109192.1 NP_001102662.1 6, 7 574
GenBank mRNA BC158681.1 - 6, 7 584

View Full Product Details