Gene Transcripts

Gene Symbol
Cpne8
Entrez Gene ID
362988
Gene Name
copine VIII
Gene Aliases
Chromosome Location
Chr. 7: 131623181 - 131798236 on Build Rnor_6.0
UniGene ID
Rn.11746
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108750.1 NP_001102220.1 18, 19 1606
GenBank mRNA AM747284.1 - 18, 19 1477
BC168840.1 - 18, 19 1529 7125016

View Full Product Details