Gene Transcripts

Gene Symbol
Fezf1
Entrez Gene ID
500048
Gene Name
Fez family zinc finger 1
Gene Aliases
RGD1560480
Chromosome Location
Chr. 4: 50309281 - 50312608 on Build Rnor_6.0
UniGene ID
Rn.218518
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109224.1 NP_001102694.1 1 883
XM_008762805.1 XP_008761027.1 2 445

View Full Product Details