Gene Transcripts

Gene Symbol
Emx1
Entrez Gene ID
500235
Gene Name
empty spiracles homeobox 1
Gene Aliases
RGD1564002
Chromosome Location
Chr. 4: 116967646 - 116984136 on Build Rnor_6.0
UniGene ID
Rn.140907
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_575584.5 XP_575584.5 3 1540
GenBank mRNA FQ212435.1 - 4 2747

View Full Product Details