Gene Transcripts

Gene Symbol
Ccdc38
Entrez Gene ID
500823
Gene Name
coiled-coil domain containing 38
Gene Aliases
RGD1564046
Chromosome Location
Chr. 7: 34402930 - 34477861 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241271.2 XP_006241333.1 17 1906
XM_008765310.1 XP_008763532.1 16 1636

View Full Product Details