Gene Transcripts

Gene Symbol
Tbc1d5
Entrez Gene ID
501088
Gene Name
TBC1 domain family, member 5
Gene Aliases
RGD1561626
Chromosome Location
Chr. 9: 1144644 - 1747732 on Build Rnor_6.0
UniGene ID
Rn.203950
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134762.1 NP_001128234.1 4 444
XM_008766768.1 XP_008764990.1 4 1500
GenBank mRNA BC162056.1 - 4 444

View Full Product Details