Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004823.1 | NP_004814.1 | 3 | 1824 | |
XM_011527526.1 | XP_011525828.1 | 4 | 2386 | ||
GenBank mRNA | AF117708.1 | - | 3 | 1824 | |
AF134149.1 | - | 3 | 1833 | ||
AK022344.1 | - | 3 | 1553 | ||
BC004367.1 | - | 3 | 1754 | 3639657 |