Gene Transcripts
- Gene Symbol
- TECR
- Entrez Gene ID
- 9524
- Gene Name
- trans-2,3-enoyl-CoA reductase
- Gene Aliases
- GPSN2, MRT14, SC2, TER
- Chromosome Location
- Chr. 19: 14527760 - 14565980 on Build GRCh38
- UniGene ID
- Hs.515642
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321170.1 |
NP_001308099.1
|
4, 5 |
314 |
|
NM_138501.5 |
NP_612510.1
|
3, 4 |
240 |
|
NR_038103.1 |
|
3, 4 |
272 |
|
NR_038104.1 |
|
3, 4 |
305 |
|
XM_006722945.1 |
XP_006723008.1
|
4, 5 |
358 |
|
XM_011528442.2 |
XP_011526744.1
|
3, 4 |
188 |
|
GenBank mRNA |
AF038958.1 |
- |
3, 4 |
186 |
|
AF222742.1 |
- |
3, 4 |
161 |
|
AK001416.1 |
- |
3, 4 |
204 |
|
AK094087.1 |
- |
4, 5 |
281 |
|
AK299134.1 |
- |
4, 5 |
278 |
|
AK315412.1 |
- |
3, 4 |
204 |
|
BC000174.2 |
- |
2, 3 |
100 |
|
BC002425.1 |
- |
3, 4 |
172 |
3163592 |
BC005952.1 |
- |
3, 4 |
183 |
4249841 |
BC007801.2 |
- |
3, 4 |
190 |
4128702 |
BC013881.2 |
- |
3, 4 |
188 |
3928698 |
BG705196.1 |
- |
3, 4 |
243 |
|
BT007179.1 |
- |
2, 3 |
110 |
|
CR456892.1 |
- |
2, 3 |
110 |
|
View Full Product Details