Gene Transcripts

Gene Symbol
Hmgn5
Entrez Gene ID
100910616
Gene Name
high mobility group nucleosome binding domain 5
Gene Aliases
Nsbp1
Chromosome Location
Chr. X: 80134557 - 80142902 on Build Rnor_6.0
UniGene ID
Rn.25395
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276434.1 NP_001263363.1 6 524
GenBank mRNA DY318327.1 - 6 423

Gene Symbol
Hmgn5b
Entrez Gene ID
681284
Gene Name
high mobility group nucleosome binding domain 5B
Gene Aliases
Hmgn5, Hmgn5l1, Nsbp1
Chromosome Location
Chr. 1: 124477164 - 124485737 on Build Rnor_6.0
UniGene ID
Rn.25395
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134706.1 NP_001128178.1 6 524
GenBank mRNA BC168165.1 - 6 426

View Full Product Details