Gene Transcripts

Gene Symbol
RAB3D
Entrez Gene ID
9545
Gene Name
RAB3D, member RAS oncogene family
Gene Aliases
D2-2, GOV, RAB16, RAD3D
Chromosome Location
Chr. 19: 11322046 - 11339668 on Build GRCh38
UniGene ID
Hs.744916
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004283.3 NP_004274.1 2 304
GenBank mRNA AF081353.1 - 2 217
AF498933.1 - 1 43
AK313538.1 - 2 304
BC016471.1 - 2 281 3861912
DA241826.1 - 2 304

View Full Product Details