Gene Transcripts

Gene Symbol
Dqx1
Entrez Gene ID
680434
Gene Name
DEAQ box RNA-dependent ATPase 1
Gene Aliases
Chromosome Location
Chr. 4: 113890198 - 113899350 on Build Rnor_6.0
UniGene ID
Rn.154059
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109407.1 NP_001102877.1 2 156
XM_006236717.2 XP_006236779.1 2 646
XM_008763030.1 XP_008761252.1 1 831

View Full Product Details