Gene Transcripts

Gene Symbol
Fam72a
Entrez Gene ID
681249
Gene Name
family with sequence similarity 72, member A
Gene Aliases
Chromosome Location
Chr. 13: 48288071 - 48298604 on Build Rnor_6.0
UniGene ID
Rn.212623
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081451.1 NP_001074920.1 2 218
GenBank mRNA AY513720.1 - 2 218

View Full Product Details