Gene Transcripts

Gene Symbol
Acyp2
Entrez Gene ID
364224
Gene Name
acylphosphatase 2, muscle type
Gene Aliases
Chromosome Location
Chr. 14: 114891245 - 115052548 on Build Rnor_6.0
UniGene ID
Rn.22481
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169145.1 NP_001162616.1 4 901
GenBank mRNA FQ215015.1 - 4 732
FQ224926.1 - 4 769

View Full Product Details