Gene Transcripts

Gene Symbol
Nrsn2
Entrez Gene ID
689978
Gene Name
neurensin 2
Gene Aliases
Chromosome Location
Chr. 3: 147843077 - 147851768 on Build Rnor_6.0
UniGene ID
Rn.138040
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109561.1 NP_001103031.1 3 342
GenBank mRNA BC159407.1 - 3 323

View Full Product Details