Gene Transcripts

Gene Symbol
Hoxd9
Entrez Gene ID
688999
Gene Name
homeo box D9
Gene Aliases
Chromosome Location
Chr. 3: 61620192 - 61622360 on Build Rnor_6.0
UniGene ID
Rn.224625
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173469.1 NP_001166940.1 2 1289
GenBank mRNA BC169092.1 - 2 1289

View Full Product Details