Gene Transcripts

Gene Symbol
Tmem29
Entrez Gene ID
100359832
Gene Name
transmembrane protein 29
Gene Aliases
Chromosome Location
Chr. X: 20821296 - 20866271 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752031.3 XP_003752079.2 3 382
XM_006256807.2 XP_006256869.1 3 5149
XM_006256808.2 XP_006256870.1 3 1235

View Full Product Details