Gene Transcripts

Gene Symbol
Trnp1
Entrez Gene ID
688990
Gene Name
TMF1-regulated nuclear protein 1
Gene Aliases
Chromosome Location
Chr. 5: 151703139 - 151710191 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001069068.5 XP_001069068.5 2 1431
XM_008764186.1 XP_008762408.1 3 1468

View Full Product Details