Gene Transcripts

Gene Symbol
Vom2r29
Entrez Gene ID
691560
Gene Name
vomeronasal 2 receptor, 29
Gene Aliases
Chromosome Location
Chr. 1: 74251167 - 74272053 on Build Rnor_6.0
UniGene ID
Rn.217633
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099496.1 NP_001092966.1 3 922

View Full Product Details