Gene Transcripts

Gene Symbol
Crtc1
Entrez Gene ID
684527
Gene Name
CREB regulated transcription coactivator 1
Gene Aliases
Mect1, TORC-1, TORC1
Chromosome Location
Chr. 16: 20740750 - 20798142 on Build Rnor_6.0
UniGene ID
Rn.208248
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047115.1 NP_001040580.1 14 1759
XM_006252936.2 XP_006252998.1 13 1748
GenBank mRNA DQ538521.1 - 14 1759

View Full Product Details