Gene Transcripts

Gene Symbol
TMEM63A
Entrez Gene ID
9725
Gene Name
transmembrane protein 63A
Gene Aliases
KIAA0792
Chromosome Location
Chr. 1: 225840556 - 225882720 on Build GRCh38
UniGene ID
Hs.119387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014698.2 NP_055513.2 7, 8 1146
XM_011544328.2 XP_011542630.1 7, 8 998
XM_011544329.2 XP_011542631.1 7, 8 2265
XM_011544330.2 XP_011542632.1 6, 7 955
XM_011544331.2 XP_011542633.1 7, 8 829
XM_017002916.1 XP_016858405.1 7, 8 800
GenBank mRNA AB018335.1 - 7, 8 774
AK022502.1 - 7, 8 795
AK222934.1 - 7, 8 776
AK222944.1 - 7, 8 776
AK309210.1 - 5, 6 723
BC030245.1 - 7, 8 779 5216414

View Full Product Details