Gene Transcripts

Gene Symbol
Mef2bnb
Entrez Gene ID
688966
Gene Name
MEF2B neighbor
Gene Aliases
Chromosome Location
Chr. 16: 21022500 - 21029163 on Build Rnor_6.0
UniGene ID
Rn.27139
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145726.1 NP_001139198.1 2 84
GenBank mRNA CB782819.1 - 2 104
DV714338.1 - 2 67

View Full Product Details