Gene Transcripts

Gene Symbol
Oc90
Entrez Gene ID
690625
Gene Name
otoconin 90
Gene Aliases
Chromosome Location
Chr. 7: 106617491 - 106697089 on Build Rnor_6.0
UniGene ID
Rn.214501
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134765.1 NP_001128237.1 14 1353
XM_008765519.1 XP_008763741.1 27 2870

View Full Product Details