Gene Transcripts

Gene Symbol
Atr
Entrez Gene ID
685055
Gene Name
ATR serine/threonine kinase
Gene Aliases
Chromosome Location
Chr. 8: 103673541 - 103772071 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750561.3 XP_003750609.1 6 1560
XM_006243600.2 XP_006243662.1 6 1951
XM_008766572.1 XP_008764794.1 6 1596

View Full Product Details