Gene Transcripts

Gene Symbol
Rprml
Entrez Gene ID
685826
Gene Name
reprimo-like
Gene Aliases
Chromosome Location
Chr. 10: 91710495 - 91711499 on Build Rnor_6.0
UniGene ID
Rn.38616
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109481.1 NP_001102951.1 1 481
GenBank mRNA FQ220063.1 - 1 511

View Full Product Details