Gene Transcripts

Gene Symbol
Tmem88b
Entrez Gene ID
680723
Gene Name
transmembrane protein 88B
Gene Aliases
Chromosome Location
Chr. 5: 173230364 - 173233188 on Build Rnor_6.0
UniGene ID
Rn.134550
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109426.1 NP_001102896.1 2 1205

View Full Product Details