Gene Transcripts

Gene Symbol
Wdr76
Entrez Gene ID
311361
Gene Name
WD repeat domain 76
Gene Aliases
RGD1309616
Chromosome Location
Chr. 3: 113445498 - 113479483 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006234874.2 XP_006234936.1 3 880
XM_006234875.2 XP_006234937.1 3 880
XM_006234876.2 XP_006234938.1 2 325
XM_006234877.1 XP_006234939.1 2 318
XM_230512.7 XP_230512.4 2 356

View Full Product Details